Assay Details
| Gene Symbol: sod-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059889.6 | NP_492290.1 | 4 - 5 | 410 | 82 | |
| GenBank mRNA | D12984.1 | - | 3 - 4 | 379 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sod-2| Gene Symbol: sod-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059889.6 | NP_492290.1 | 4 - 5 | 410 | 82 | |
| GenBank mRNA | D12984.1 | - | 3 - 4 | 379 | 82 | |
Set Membership: |
|