Assay Details
| Gene Symbol: axl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047206.5 | NP_001040671.1 | 6 - 7 | 710 | 84 | |
| NM_001047207.6 | NP_001040672.1 | 7 - 8 | 831 | 84 | ||
| GenBank mRNA | DQ185512.1 | - | 7 - 8 | 830 | 84 | |
More Information
Set Membership: |
|

axl-1
Genomic Map