Assay Details
| Gene Symbol: gon-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129034.3 | NP_001122506.1 | 14 - 15 | 3492 | 97 | |
| NM_001136316.4 | NP_001129788.1 | 8 - 9 | 2457 | 97 | ||
| NM_001373178.2 | NP_001359707.1 | 16 - 17 | 3643 | 97 | ||
| NM_059914.7 | NP_492315.4 | 17 - 18 | 3716 | 97 | ||
More Information
Set Membership: |
|

gon-2
Genomic Map