Assay Details
| Gene Symbol: gon-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129034.3 | NP_001122506.1 | 18 - 19 | 4015 | 132 | |
| NM_001136316.4 | NP_001129788.1 | 12 - 13 | 2980 | 132 | ||
| NM_001373178.2 | NP_001359707.1 | 20 - 21 | 4166 | 132 | ||
| NM_059914.7 | NP_492315.4 | 21 - 22 | 4239 | 132 | ||
More Information
Set Membership: |
|

gon-2
Genomic Map