Assay Details
| Gene Symbol: gon-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129034.3 | NP_001122506.1 | 19 - 20 | 4325 | 115 | |
| NM_001136316.4 | NP_001129788.1 | 13 - 14 | 3290 | 115 | ||
| NM_001373178.2 | NP_001359707.1 | 21 - 22 | 4476 | 115 | ||
| NM_059914.7 | NP_492315.4 | 22 - 23 | 4549 | 115 | ||
More Information
Set Membership: |
|

gon-2
Genomic Map