Assay Details
| Gene Symbol: gon-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129034.3 | NP_001122506.1 | 21 - 22 | 4544 | 77 | |
| NM_001136316.4 | NP_001129788.1 | 15 - 16 | 3509 | 77 | ||
| NM_001373178.2 | NP_001359707.1 | 23 - 24 | 4695 | 77 | ||
| NM_059914.7 | NP_492315.4 | 24 - 25 | 4768 | 77 | ||
More Information
Set Membership: |
|

gon-2
Genomic Map