Assay Details
| Gene Symbol: gon-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129033.3 | NP_001122505.1 | 4 - 5 | 299 | 121 | |
| NM_001129034.3 | NP_001122506.1 | 1 - 2 | 116 | 121 | ||
| NM_001373178.2 | NP_001359707.1 | 3 - 4 | 267 | 121 | ||
| NM_059914.7 | NP_492315.4 | 4 - 5 | 340 | 121 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001373178.2 transcripts.
Set Membership: |
|

gon-2
Genomic Map