Assay Details
| Gene Symbol: gon-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129033.3 | NP_001122505.1 | 5 - 6 | 416 | 90 | |
| NM_001129034.3 | NP_001122506.1 | 2 - 3 | 233 | 90 | ||
| NM_001373178.2 | NP_001359707.1 | 4 - 5 | 384 | 90 | ||
| NM_059914.7 | NP_492315.4 | 5 - 6 | 457 | 90 | ||
More Information
Set Membership: |
|

gon-2
Genomic Map