Assay Details
| Gene Symbol: gon-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129034.3 | NP_001122506.1 | 5 - 6 | 891 | 122 | |
| NM_001373178.2 | NP_001359707.1 | 7 - 8 | 1042 | 122 | ||
| NM_059914.7 | NP_492315.4 | 8 - 9 | 1115 | 122 | ||
More Information
Set Membership: |
|

gon-2
Genomic Map