Assay Details
| Gene Symbol: gon-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129034.3 | NP_001122506.1 | 6 - 7 | 970 | 98 | |
| NM_001373178.2 | NP_001359707.1 | 8 - 9 | 1121 | 98 | ||
| NM_059914.7 | NP_492315.4 | 9 - 10 | 1194 | 98 | ||
More Information
Set Membership: |
|

gon-2
Genomic Map