Assay Details
| Gene Symbol: gon-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129034.3 | NP_001122506.1 | 7 - 8 | 1113 | 89 | |
| NM_001136316.4 | NP_001129788.1 | 1 - 2 | 78 | 89 | ||
| NM_001373178.2 | NP_001359707.1 | 9 - 10 | 1264 | 89 | ||
| NM_059914.7 | NP_492315.4 | 10 - 11 | 1337 | 89 | ||
More Information
Set Membership: |
|

gon-2
Genomic Map