Assay Details
| Gene Symbol: ran-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059921.5 | NP_492322.1 | 3 - 4 | 296 | 91 | |
| Gene Symbol: tmem-208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059922.6 | NP_492323.1 | 2 - 3 | 341 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_059922.6 transcripts.
Set Membership: |
|

ran-4
Genomic Map