Assay Details
| Gene Symbol: ddx-52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059925.6 | NP_492326.1 | 4 - 5 | 652 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ddx-52| Gene Symbol: ddx-52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059925.6 | NP_492326.1 | 4 - 5 | 652 | 85 | |
Set Membership: |
|