Assay Details
Gene Symbol: lmd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129003.3 | NP_001122475.1 | 2 - 3 | 336 | 85 |
Gene Symbol: F43G9.13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059928.5 | NP_492329.1 | 7 - 8 | 1929 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
