Assay Details
Gene Symbol: lmd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129003.3 | NP_001122475.1 | 3 - 4 | 429 | 84 | |
NM_001129004.3 | NP_001122476.1 | 3 - 4 | 437 | 84 |
Gene Symbol: F43G9.13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059928.5 | NP_492329.1 | 8 - 9 | 2022 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
