Assay Details
| Gene Symbol: fer-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129005.4 | NP_001122477.1 | 8 - 9 | 2370 | 73 | |
| NM_059936.4 | NP_492337.1 | 10 - 11 | 2769 | 73 | ||
More Information
Set Membership: |
|

fer-1
Genomic Map