Assay Details
| Gene Symbol: fer-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129005.4 | NP_001122477.1 | 11 - 12 | 3345 | 94 | |
| NM_059936.4 | NP_492337.1 | 13 - 14 | 3744 | 94 | ||
More Information
Set Membership: |
|

fer-1
Genomic Map