Assay Details
| Gene Symbol: fer-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129005.4 | NP_001122477.1 | 18 - 19 | 5520 | 72 | |
| NM_059936.4 | NP_492337.1 | 20 - 21 | 5919 | 72 | ||
More Information
Set Membership: |
|

fer-1
Genomic Map