Assay Details
| Gene Symbol: egg-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059947.6 | NP_492348.1 | 1 - 2 | 189 | 71 | |
| GenBank mRNA | JQ292908.1 | - | 1 - 2 | 101 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

egg-6| Gene Symbol: egg-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059947.6 | NP_492348.1 | 1 - 2 | 189 | 71 | |
| GenBank mRNA | JQ292908.1 | - | 1 - 2 | 101 | 71 | |
Set Membership: |
|