Assay Details
| Gene Symbol: egg-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059947.6 | NP_492348.1 | 7 - 8 | 1738 | 71 | |
| GenBank mRNA | JQ292908.1 | - | 7 - 8 | 1650 | 71 | |
More Information
Set Membership: |
|

egg-6
Genomic Map