Assay Details
| Gene Symbol: mrps-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059950.5 | NP_492351.1 | 7 - 8 | 756 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mrps-15| Gene Symbol: mrps-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059950.5 | NP_492351.1 | 7 - 8 | 756 | 95 | |
Set Membership: |
|