Assay Details
| Gene Symbol: dhfr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059963.6 | NP_492364.1 | 2 - 3 | 218 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dhfr-1| Gene Symbol: dhfr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059963.6 | NP_492364.1 | 2 - 3 | 218 | 78 | |
Set Membership: |
|