Assay Details
| Gene Symbol: gls-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001263949.3 | NP_001250878.1 | 13 - 14 | 2329 | 61 | |
| NM_059964.4 | NP_492365.3 | 13 - 14 | 2393 | 61 | ||
| NM_182082.6 | NP_871882.2 | 13 - 14 | 2393 | 61 | ||
More Information
Set Membership: |
|

gls-1
Genomic Map