Assay Details
| Gene Symbol: imp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025852.5 | NP_001021023.1 | 6 - 7 | 954 | 74 | |
| NM_001025853.5 | NP_001021024.1 | 6 - 7 | 954 | 74 | ||
| NM_001025854.6 | NP_001021025.1 | 6 - 7 | 961 | 74 | ||
More Information
Set Membership: |
|

imp-1
Genomic Map