Assay Details
| Gene Symbol: madd-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001264004.4 | NP_001250933.1 | 4 - 5 | 591 | 68 | |
| NM_060004.5 | NP_492405.3 | 4 - 5 | 586 | 68 | ||
More Information
Set Membership: |
|

madd-4
Genomic Map