Assay Details
| Gene Symbol: gld-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060045.6 | NP_492446.3 | 1 - 2 | 156 | 69 | |
| GenBank mRNA | FJ610054.1 | - | 2 - 3 | 178 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gld-4| Gene Symbol: gld-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060045.6 | NP_492446.3 | 1 - 2 | 156 | 69 | |
| GenBank mRNA | FJ610054.1 | - | 2 - 3 | 178 | 69 | |
Set Membership: |
|