Assay Details
| Gene Symbol: pyk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026241.6 | NP_001021412.1 | 2 - 3 | 157 | 70 | |
| NM_001264085.4 | NP_001251014.1 | 3 - 4 | 1261 | 70 | ||
| NM_060057.6 | NP_492458.1 | 2 - 3 | 169 | 70 | ||
More Information
Set Membership: |
|

pyk-1
Genomic Map