Assay Details
| Gene Symbol: pyk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026241.6 | NP_001021412.1 | 4 - 5 | 1043 | 63 | |
| NM_001264085.4 | NP_001251014.1 | 5 - 6 | 2147 | 63 | ||
| NM_001383589.1 | 5 - 6 | 1532 | 63 | |||
| NM_060057.6 | NP_492458.1 | 4 - 5 | 1055 | 63 | ||
| NM_060058.5 | NP_492459.1 | 4 - 5 | 1168 | 63 | ||
More Information
Set Membership: |
|

pyk-1
Genomic Map