Assay Details
| Gene Symbol: tat-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026286.5 | NP_001021457.1 | 2 - 3 | 441 | 115 | |
| NM_001026287.6 | NP_001021458.2 | 3 - 4 | 348 | 115 | ||
| NM_001047200.3 | NP_001040665.1 | 2 - 3 | 432 | 115 | ||
More Information
Set Membership: |
|

tat-5
Genomic Map