Assay Details
| Gene Symbol: tat-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026286.5 | NP_001021457.1 | 7 - 8 | 2704 | 127 | |
| NM_001026287.6 | NP_001021458.2 | 8 - 9 | 2611 | 127 | ||
| NM_001047200.3 | NP_001040665.1 | 7 - 8 | 2695 | 127 | ||
More Information
Set Membership: |
|

tat-5
Genomic Map