Assay Details
| Gene Symbol: F36H2.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348615.3 | NP_001335526.1 | 14 | 2824 | 135 | |
| NM_001348616.3 | NP_001335525.1 | 14 | 2824 | 135 | ||
| NM_001373341.1 | NP_001359596.1 | 13 - 14 | 2817 | 135 | ||
| NM_001373348.2 | NP_001360153.1 | 15 | 2853 | 135 | ||
| NM_001381014.1 | NP_001368097.1 | 11 - 12 | 2191 | 135 | ||
| NM_001381017.1 | NP_001368096.1 | 12 - 13 | 2401 | 135 | ||
More Information
Set Membership: |
|

F36H2.3
Genomic Map