Assay Details
| Gene Symbol: F36H2.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001264110.3 | NP_001251039.1 | 12 - 13 | 2813 | 92 | |
| NM_001264112.3 | NP_001251041.1 | 14 - 15 | 3239 | 92 | ||
| NM_001264114.3 | NP_001251043.1 | 15 - 16 | 3446 | 92 | ||
| NM_001348615.3 | NP_001335526.1 | 17 | 3869 | 92 | ||
| NM_001348616.3 | NP_001335525.1 | 17 | 3869 | 92 | ||
| NM_001373341.1 | NP_001359596.1 | 16 - 17 | 3862 | 92 | ||
| NM_001373348.2 | NP_001360153.1 | 18 | 3898 | 92 | ||
| NM_001381014.1 | NP_001368097.1 | 14 - 15 | 3236 | 92 | ||
| NM_001381017.1 | NP_001368096.1 | 15 - 16 | 3446 | 92 | ||
More Information
Set Membership: |
|

F36H2.3
Genomic Map