Assay Details
| Gene Symbol: F36H2.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001264110.3 | NP_001251039.1 | 13 - 14 | 2931 | 79 | |
| NM_001264112.3 | NP_001251041.1 | 15 - 16 | 3357 | 79 | ||
| NM_001264114.3 | NP_001251043.1 | 16 - 17 | 3564 | 79 | ||
| NM_001373341.1 | NP_001359596.1 | 17 - 18 | 3980 | 79 | ||
| NM_001373348.2 | NP_001360153.1 | 18 - 19 | 4016 | 79 | ||
| NM_001381014.1 | NP_001368097.1 | 15 - 16 | 3354 | 79 | ||
| NM_001381017.1 | NP_001368096.1 | 16 - 17 | 3564 | 79 | ||
More Information
Set Membership: |
|

F36H2.3
Genomic Map