Assay Details
| Gene Symbol: F36H2.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348616.3 | NP_001335525.1 | 20 | 4288 | 70 | |
| NM_001373341.1 | NP_001359596.1 | 19 - 20 | 4680 | 70 | ||
More Information
Set Membership: |
|

F36H2.3
Genomic Map