Assay Details
| Gene Symbol: F36H2.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348615.3 | NP_001335526.1 | 19 | 4061 | 71 | |
| NM_001348616.3 | NP_001335525.1 | 22 | 4481 | 71 | ||
| NM_001373341.1 | NP_001359596.1 | 21 - 22 | 4873 | 71 | ||
| NM_001373348.2 | NP_001360153.1 | 22 | 4953 | 71 | ||
More Information
Set Membership: |
|

F36H2.3
Genomic Map