Assay Details
| Gene Symbol: F36H2.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001264110.3 | NP_001251039.1 | 8 - 9 | 1558 | 93 | |
| NM_001264112.3 | NP_001251041.1 | 8 - 9 | 1558 | 93 | ||
| NM_001264114.3 | NP_001251043.1 | 8 - 9 | 1558 | 93 | ||
| NM_001348615.3 | NP_001335526.1 | 8 | 1558 | 93 | ||
| NM_001348616.3 | NP_001335525.1 | 8 | 1558 | 93 | ||
| NM_001373341.1 | NP_001359596.1 | 7 - 8 | 1551 | 93 | ||
| NM_001373348.2 | NP_001360153.1 | 9 | 1587 | 93 | ||
| NM_001381014.1 | NP_001368097.1 | 8 - 9 | 1558 | 93 | ||
| NM_001381017.1 | NP_001368096.1 | 8 - 9 | 1558 | 93 | ||
More Information
Set Membership: |
|

F36H2.3
Genomic Map