Assay Details
| Gene Symbol: F36H2.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001264114.3 | NP_001251043.1 | 10 - 11 | 1978 | 138 | |
| NM_001348615.3 | NP_001335526.1 | 10 | 1978 | 138 | ||
| NM_001348616.3 | NP_001335525.1 | 10 | 1978 | 138 | ||
| NM_001373341.1 | NP_001359596.1 | 9 - 10 | 1971 | 138 | ||
| NM_001373348.2 | NP_001360153.1 | 11 | 2007 | 138 | ||
More Information
Set Membership: |
|

F36H2.3
Genomic Map