Assay Details
| Gene Symbol: F26H9.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060077.6 | NP_492478.2 | 9 - 10 | 1538 | 134 | |
| Gene Symbol: rpoa-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060075.6 | NP_492476.1 | 3 - 4 | 667 | 134 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_060075.6 transcripts.
Set Membership: |
|

F26H9.2
Genomic Map