Assay Details
| Gene Symbol: rtcb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060097.5 | NP_492498.1 | 1 - 2 | 250 | 87 | |
| NR_136370.1 | 1 - 2 | 249 | 87 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rtcb-1| Gene Symbol: rtcb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060097.5 | NP_492498.1 | 1 - 2 | 250 | 87 | |
| NR_136370.1 | 1 - 2 | 249 | 87 | |||
Set Membership: |
|