Assay Details
| Gene Symbol: stl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001264176.3 | NP_001251105.1 | 1 - 2 | 55 | 89 | |
| NM_001264177.3 | NP_001251106.1 | 2 - 3 | 197 | 89 | ||
More Information
Set Membership: |
|

stl-1
Genomic Map