Assay Details
| Gene Symbol: stl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001264176.3 | NP_001251105.1 | 2 - 3 | 191 | 132 | |
| NM_001264177.3 | NP_001251106.1 | 3 - 4 | 333 | 132 | ||
More Information
Set Membership: |
|

stl-1
Genomic Map