Assay Details
| Gene Symbol: stl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001264176.3 | NP_001251105.1 | 4 - 5 | 654 | 78 | |
| NM_001264177.3 | NP_001251106.1 | 5 - 6 | 796 | 78 | ||
More Information
Set Membership: |
|

stl-1
Genomic Map