Assay Details
| Gene Symbol: usp-48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375101.1 | NP_001361851.1 | 4 - 5 | 828 | 147 | |
| NM_001375102.2 | NP_001362163.1 | 4 - 5 | 834 | 147 | ||
More Information
Set Membership: |
|

usp-48
Genomic Map