Assay Details
| Gene Symbol: usp-48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375102.2 | NP_001362163.1 | 7 - 8 | 2529 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

usp-48| Gene Symbol: usp-48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375102.2 | NP_001362163.1 | 7 - 8 | 2529 | 88 | |
Set Membership: |
|