Assay Details
| Gene Symbol: ahr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025865.4 | NP_001021036.1 | 6 - 7 | 836 | 56 | |
| NM_001025866.5 | NP_001021037.1 | 5 - 6 | 770 | 56 | ||
| GenBank mRNA | AF039570.1 | - | 6 - 7 | 832 | 56 | |
More Information
Set Membership: |
|

ahr-1
Genomic Map