Assay Details
| Gene Symbol: ahr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025865.4 | NP_001021036.1 | 8 - 9 | 1044 | 151 | |
| NM_001025866.5 | NP_001021037.1 | 7 - 8 | 978 | 151 | ||
| GenBank mRNA | AF039570.1 | - | 8 - 9 | 1040 | 151 | |
More Information
Set Membership: |
|

ahr-1
Genomic Map