Assay Details
| Gene Symbol: ikb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060174.6 | NP_492575.1 | 1 - 2 | 228 | 90 | |
| GenBank mRNA | AF348169.1 | - | 1 - 2 | 72 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ikb-1| Gene Symbol: ikb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060174.6 | NP_492575.1 | 1 - 2 | 228 | 90 | |
| GenBank mRNA | AF348169.1 | - | 1 - 2 | 72 | 90 | |
Set Membership: |
|