Assay Details
| Gene Symbol: ikb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060174.6 | NP_492575.1 | 5 - 6 | 768 | 118 | |
| GenBank mRNA | AF348169.1 | - | 5 - 6 | 612 | 118 | |
More Information
Set Membership: |
|

ikb-1
Genomic Map