Assay Details
| Gene Symbol: ikb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060174.6 | NP_492575.1 | 6 - 7 | 895 | 146 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ikb-1| Gene Symbol: ikb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060174.6 | NP_492575.1 | 6 - 7 | 895 | 146 | |
Set Membership: |
|