Assay Details
| Gene Symbol: ikb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060174.6 | NP_492575.1 | 7 - 8 | 1164 | 98 | |
| GenBank mRNA | AF348169.1 | - | 7 - 8 | 1008 | 98 | |
More Information
Set Membership: |
|

ikb-1
Genomic Map